Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAATAATTAGTTCTGCTCTTAT[A/G]TAATGTTATCTGTTAGGCTTTATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC81 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC81 - coiled-coil domain containing 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001156474.1 | Intron | NP_001149946.1 | ||||
NM_021827.4 | Intron | NP_068599.3 | ||||
XM_005274161.1 | Intron | XP_005274218.1 | ||||
XM_005274162.3 | Intron | XP_005274219.1 | ||||
XM_017018105.1 | Intron | XP_016873594.1 |