Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGGTAGTTAAGTTTCTTGGTATC[A/C]CAGTAGTACTTCTTATTATCCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603441 | ||||||||||||||||||||
Literature Links: |
ATP5S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5S - ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKL1 - cyclin dependent kinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282236.1 | 1404 | Intron | NP_001269165.1 | |||
NM_004196.4 | 1404 | Missense Mutation | TGG,TGT | W,C 346 | NP_004187.2 | |
XM_005268157.2 | 1404 | Missense Mutation | TGG,TGT | W,C 346 | XP_005268214.1 | |
XM_005268159.2 | 1404 | Missense Mutation | TGG,TGT | W,C 289 | XP_005268216.1 | |
XM_005268160.4 | 1404 | Missense Mutation | TGG,TGT | W,C 232 | XP_005268217.1 | |
XM_011537276.1 | 1404 | Missense Mutation | TGG,TGT | W,C 296 | XP_011535578.1 | |
XM_017021729.1 | 1404 | Missense Mutation | TGG,TGT | W,C 346 | XP_016877218.1 | |
XM_017021730.1 | 1404 | Missense Mutation | TGG,TGT | W,C 289 | XP_016877219.1 | |
XM_017021731.1 | 1404 | Missense Mutation | TGG,TGT | W,C 289 | XP_016877220.1 | |
XM_017021732.1 | 1404 | UTR 3 | XP_016877221.1 |