Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGTTTTGGCTTAAGCTGAGGAG[A/T]CTGCTTTCTTAAGTATGGTCTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615260 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01599 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC01599 - long intergenic non-protein coding RNA 1599 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VCPKMT - valosin containing protein lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040662.1 | 6556 | Intron | NP_001035752.1 | |||
NM_024558.2 | 6556 | Intron | NP_078834.2 | |||
XM_005268060.3 | 6556 | Intron | XP_005268117.1 | |||
XM_017021640.1 | 6556 | UTR 3 | XP_016877129.1 | |||
XM_017021641.1 | 6556 | UTR 3 | XP_016877130.1 | |||
XM_017021642.1 | 6556 | Intron | XP_016877131.1 |