Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603749 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRPM2 - transient receptor potential cation channel subfamily M member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320350.1 | Intron | NP_001307279.1 | ||||
NM_001320351.1 | Intron | NP_001307280.1 | ||||
NM_001320352.1 | Intron | NP_001307281.1 | ||||
NM_003307.3 | Intron | NP_003298.1 | ||||
XM_005261171.3 | Intron | XP_005261228.1 | ||||
XM_011529736.2 | Intron | XP_011528038.1 | ||||
XM_017028456.1 | Intron | XP_016883945.1 | ||||
XM_017028457.1 | Intron | XP_016883946.1 |
Set Membership: |
HapMap |