Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGAAAAGAATTATGAAGACACA[G/T]CACTGAATATTCACTAGTTACTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604554 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSF2BP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HSF2BP - heat shock transcription factor 2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007031.1 | Intron | NP_008962.1 | ||||
XM_011529443.2 | Intron | XP_011527745.2 | ||||
XM_011529445.2 | Intron | XP_011527747.2 | ||||
XM_011529446.2 | Intron | XP_011527748.1 | ||||
XM_011529447.2 | Intron | XP_011527749.1 | ||||
XM_017028267.1 | Intron | XP_016883756.1 | ||||
XM_017028268.1 | Intron | XP_016883757.1 | ||||
XM_017028269.1 | Intron | XP_016883758.1 | ||||
XM_017028270.1 | Intron | XP_016883759.1 | ||||
XM_017028271.1 | Intron | XP_016883760.1 | ||||
XM_017028272.1 | Intron | XP_016883761.1 |