Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCAGACACACCAGGGGGCCTCC[C/T]GAATCGCCCTGCAAAGGAGATAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610469 MIM: 173370 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3M2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP3M2 - adaptor related protein complex 3 mu 2 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAT - plasminogen activator, tissue type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000930.4 | 1658 | Silent Mutation | TCA,TCG | S,S 513 | NP_000921.1 | |
NM_001319189.1 | 1658 | Silent Mutation | TCA,TCG | S,S 424 | NP_001306118.1 | |
NM_033011.3 | 1658 | Silent Mutation | TCA,TCG | S,S 467 | NP_127509.1 |
Set Membership: |
HapMap |