Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 174760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLB - DNA polymerase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002690.2 | Intron | NP_002681.1 | ||||
XM_005273535.3 | Intron | XP_005273592.1 | ||||
XM_005273536.3 | Intron | XP_005273593.1 | ||||
XM_005273537.3 | Intron | XP_005273594.1 | ||||
XM_005273538.2 | Intron | XP_005273595.1 | ||||
XM_005273539.2 | Intron | XP_005273596.1 | ||||
XM_005273540.4 | Intron | XP_005273597.1 | ||||
XM_006716353.2 | Intron | XP_006716416.1 | ||||
XM_017013583.1 | Intron | XP_016869072.1 | ||||
XM_017013584.1 | Intron | XP_016869073.1 |