Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTGTTTTGTTTTGGCAGTCTG[A/C]CATAACTTTATACTAATGCAGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 143024 | ||||||||||||||||||||
Literature Links: |
GNL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNL1 - G protein nucleolar 1 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005275.3 | 6072 | UTR 3 | NP_005266.2 | |||
XM_005249015.3 | 6072 | UTR 3 | XP_005249072.1 |