Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGATTGCTTCTGTCTGTTATTTA[G/T]ATATGGAAGCTGAGGGGATGCACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611685 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF8 - ring finger protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003958.3 | 105 | UTR 5 | NP_003949.1 | |||
NM_183078.2 | 105 | UTR 5 | NP_898901.1 | |||
XM_006715241.2 | 105 | UTR 5 | XP_006715304.1 | |||
XM_006715242.2 | 105 | UTR 5 | XP_006715305.1 | |||
XM_017011462.1 | 105 | Intron | XP_016866951.1 | |||
XM_017011463.1 | 105 | Intron | XP_016866952.1 | |||
XM_017011464.1 | 105 | UTR 5 | XP_016866953.1 |
Set Membership: |
HapMap |