Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGGATGAGAGGCGTAGGTGAA[A/G]AGAAAGGGGTCTTCTTGAGGTGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603715 | ||||||||||||||||||||
Literature Links: |
GCM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCM1 - glial cells missing homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003643.3 | 1379 | Silent Mutation | CTC,CTT | L,L 390 | NP_003634.2 | |
XM_017011390.1 | 1379 | Silent Mutation | CTC,CTT | L,L 452 | XP_016866879.1 |