Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCACTAGGTGACTAGGGAGGCTG[C/G]GCCTGATTTGGCTCACTCGTGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UNC5B - unc-5 netrin receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244889.1 | Intron | NP_001231818.1 | ||||
NM_170744.4 | Intron | NP_734465.2 | ||||
XM_011539453.1 | Intron | XP_011537755.1 | ||||
XM_017015834.1 | Intron | XP_016871323.1 | ||||
XM_017015835.1 | Intron | XP_016871324.1 |
UNC5B-AS1 - UNC5B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |