Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAATAAGTTTGGAGCTTCCACT[G/T]TGAGCCGGGCACTAGGGACACGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610045 MIM: 602515 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH5A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH5A1 - aldehyde dehydrogenase 5 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080.3 | Intron | NP_001071.1 | ||||
NM_170740.1 | Intron | NP_733936.1 |
GPLD1 - glycosylphosphatidylinositol specific phospholipase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001503.3 | Intron | NP_001494.2 | ||||
XM_011514509.2 | Intron | XP_011512811.1 | ||||
XM_017010753.1 | Intron | XP_016866242.1 | ||||
XM_017010754.1 | Intron | XP_016866243.1 |