Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCCCGCGCCAGGCTCGGCTCT[C/T]AGTCCAGCAGGCGTGTGCGGTGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116949 MIM: 117140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC25B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC25B - cell division cycle 25B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287516.1 | 835 | Intron | NP_001274445.1 | |||
NM_001287517.1 | 835 | Intron | NP_001274446.1 | |||
NM_001287518.1 | 835 | Intron | NP_001274447.1 | |||
NM_001287519.1 | 835 | Intron | NP_001274448.1 | |||
NM_001287520.1 | 835 | Intron | NP_001274449.1 | |||
NM_001287522.1 | 835 | Intron | NP_001274451.1 | |||
NM_001287524.1 | 835 | Intron | NP_001274453.1 | |||
NM_004358.4 | 835 | Silent Mutation | CTC,CTT | L,L 14 | NP_004349.1 | |
NM_021872.3 | 835 | Silent Mutation | CTC,CTT | L,L 14 | NP_068658.1 | |
NM_021873.3 | 835 | Silent Mutation | CTC,CTT | L,L 14 | NP_068659.1 |
CENPB - centromere protein B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |