Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTTAAGGTTGGCTCAGAGATT[C/T]TGGGAAGAAATAATGTATGGTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602414 | ||||||||||||||||||||
Literature Links: |
APBA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APBA1 - amyloid beta precursor protein binding family A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163.3 | 3561 | UTR 3 | NP_001154.2 | |||
XM_005251968.3 | 3561 | UTR 3 | XP_005252025.1 | |||
XM_011518617.2 | 3561 | UTR 3 | XP_011516919.1 | |||
XM_017014670.1 | 3561 | UTR 3 | XP_016870159.1 |