Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTAGGGAACCTGCCTAATGTCAC[A/G]CAGCAATTGAATCAGAGGCAGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611420 MIM: 602377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIZ1 - CDKN1A interacting zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNM1 - dynamin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005336.2 | Intron | NP_001005336.1 | ||||
NM_001288737.1 | Intron | NP_001275666.1 | ||||
NM_001288738.1 | Intron | NP_001275667.1 | ||||
NM_001288739.1 | Intron | NP_001275668.1 | ||||
NM_004408.3 | Intron | NP_004399.2 | ||||
XM_005251763.2 | Intron | XP_005251820.1 | ||||
XM_005251768.2 | Intron | XP_005251825.1 | ||||
XM_005251769.2 | Intron | XP_005251826.1 | ||||
XM_006716992.2 | Intron | XP_006717055.1 | ||||
XM_006716993.1 | Intron | XP_006717056.1 | ||||
XM_011518335.2 | Intron | XP_011516637.1 | ||||
XM_011518336.2 | Intron | XP_011516638.1 | ||||
XM_017014369.1 | Intron | XP_016869858.1 | ||||
XM_017014370.1 | Intron | XP_016869859.1 | ||||
XM_017014371.1 | Intron | XP_016869860.1 | ||||
XM_017014372.1 | Intron | XP_016869861.1 | ||||
XM_017014373.1 | Intron | XP_016869862.1 |