Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCCTTAAAGTCGAGGTACTTTT[C/T]ATCTTTCAAAATGCATTCGTTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616626 | ||||||||||||||||||||
Literature Links: |
CERCAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CERCAM - cerebral endothelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286760.1 | 691 | Intron | NP_001273689.1 | |||
NM_016174.4 | 691 | Intron | NP_057258.3 | |||
XM_005252035.1 | 691 | Intron | XP_005252092.1 | |||
XM_011518762.1 | 691 | Intron | XP_011517064.1 | |||
XM_011518763.2 | 691 | UTR 5 | XP_011517065.1 | |||
XM_017014794.1 | 691 | Intron | XP_016870283.1 |