Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603371 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLE1 - GLE1, RNA export mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003722.1 | Intron | NP_001003722.1 | ||||
NM_001499.2 | Intron | NP_001490.1 | ||||
XM_006717059.2 | Intron | XP_006717122.1 | ||||
XM_006717060.2 | Intron | XP_006717123.1 | ||||
XM_011518549.1 | Intron | XP_011516851.1 | ||||
XM_011518550.1 | Intron | XP_011516852.1 | ||||
XM_011518551.1 | Intron | XP_011516853.1 |
Set Membership: |
HapMap |