Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCCTGGGTGAGTACTGGAGTTC[C/T]AAAGACACGTGAGCTAACCCTGCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 168820 MIM: 602468 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PON1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PON1 - paraoxonase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000446.5 | 1575 | UTR 3 | NP_000437.3 |
PPP1R9A - protein phosphatase 1 regulatory subunit 9A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |