Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATGCATTTGAGGTTAGGTCTTC[G/T]CAACCATGAAAACACAGTTATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPR2 - inositol 1,4,5-trisphosphate receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002223.3 | 8760 | UTR 3 | NP_002214.2 | |||
XM_017019266.1 | 8760 | UTR 3 | XP_016874755.1 | |||
XM_017019267.1 | 8760 | UTR 3 | XP_016874756.1 | |||
XM_017019268.1 | 8760 | UTR 3 | XP_016874757.1 | |||
XM_017019269.1 | 8760 | Intron | XP_016874758.1 |