Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615079 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018164.2 | Intron | NP_060634.2 | ||||
XM_005253422.3 | Intron | XP_005253479.1 | ||||
XM_011520750.2 | Intron | XP_011519052.1 | ||||
XM_011520751.2 | Intron | XP_011519053.1 | ||||
XM_011520752.2 | Intron | XP_011519054.1 | ||||
XM_011520753.2 | Intron | XP_011519055.1 | ||||
XM_017019632.1 | Intron | XP_016875121.1 | ||||
XM_017019633.1 | Intron | XP_016875122.1 | ||||
XM_017019634.1 | Intron | XP_016875123.1 | ||||
XM_017019635.1 | Intron | XP_016875124.1 | ||||
XM_017019636.1 | Intron | XP_016875125.1 | ||||
XM_017019637.1 | Intron | XP_016875126.1 |