Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATACTCAGACAAGCTATCAAACA[A/G]ATAGAGGGAGCCAGCCATTTTTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616906 MIM: 190070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CASC1 - cancer susceptibility candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRAS - KRAS proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM5 - LYR motif containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001660.2 | Intron | NP_001001660.2 | ||||
XM_005253319.4 | Intron | XP_005253376.1 | ||||
XM_005253320.4 | Intron | XP_005253377.1 | ||||
XM_017018850.1 | Intron | XP_016874339.1 |