Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTATAGTTGTATATACATTTA[A/G]GATGCTTATGTCTTCTTGGTGATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615079 MIM: 608858 | ||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FGFR1OP2 - FGFR1 oncogene partner 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171887.1 | Intron | NP_001165358.1 | ||||
NM_001171888.1 | Intron | NP_001165359.1 | ||||
NM_015633.2 | Intron | NP_056448.1 |