Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGAAGCAAAGTGTTACATCAT[C/T]GAAATGTATAAGTTTCAACTATTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608858 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR1OP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGFR1OP2 - FGFR1 oncogene partner 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171887.1 | Intron | NP_001165358.1 | ||||
NM_001171888.1 | Intron | NP_001165359.1 | ||||
NM_015633.2 | Intron | NP_056448.1 |