Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGAAAAAGACTATTCTTACTGT[C/G]TCCAGAATTGGTTCCTTCCAGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC91 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC91 - coiled-coil domain containing 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253413.1 | Intron | XP_005253470.1 | ||||
XM_005253414.1 | Intron | XP_005253471.1 | ||||
XM_005253415.1 | Intron | XP_005253472.1 | ||||
XM_006719103.3 | Intron | XP_006719166.1 | ||||
XM_006719104.2 | Intron | XP_006719167.1 | ||||
XM_006719105.1 | Intron | XP_006719168.1 | ||||
XM_011520729.2 | Intron | XP_011519031.1 | ||||
XM_011520731.2 | Intron | XP_011519033.1 | ||||
XM_011520733.2 | Intron | XP_011519035.1 | ||||
XM_011520734.1 | Intron | XP_011519036.1 | ||||
XM_011520735.1 | Intron | XP_011519037.1 | ||||
XM_017019569.1 | Intron | XP_016875058.1 | ||||
XM_017019570.1 | Intron | XP_016875059.1 | ||||
XM_017019571.1 | Intron | XP_016875060.1 | ||||
XM_017019572.1 | Intron | XP_016875061.1 | ||||
XM_017019573.1 | Intron | XP_016875062.1 | ||||
XM_017019574.1 | Intron | XP_016875063.1 | ||||
XM_017019575.1 | Intron | XP_016875064.1 | ||||
XM_017019576.1 | Intron | XP_016875065.1 | ||||
XM_017019577.1 | Intron | XP_016875066.1 |
LOC729291 - translation initiation factor IF-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |