Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTCAAATTTTAGCACATAAGTT[A/G]AAGTTTAGTAGTAAGTTAACTACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611032 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPATA17 - spermatogenesis associated 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138796.3 | Intron | NP_620151.1 | ||||
XM_005273052.1 | Intron | XP_005273109.1 | ||||
XM_006711165.3 | Intron | XP_006711228.1 | ||||
XM_011509194.2 | Intron | XP_011507496.1 |