Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGATTGGAAGCCTCTTCTATTT[G/T]CAAACATTCAGGAATGGAAATTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605484 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCAMR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCAMR - Fc fragment of IgA and IgM receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122979.2 | 2234 | UTR 3 | NP_001116451.1 | |||
NM_001170631.1 | 2234 | UTR 3 | NP_001164102.1 | |||
NM_032029.4 | 2234 | UTR 3 | NP_114418.2 | |||
XM_011510051.2 | 2234 | UTR 3 | XP_011508353.1 | |||
XM_011510052.2 | 2234 | UTR 3 | XP_011508354.1 | |||
XM_017002442.1 | 2234 | UTR 3 | XP_016857931.1 |
Set Membership: |
HapMap |