Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGCTGTGTCTTTAAGCAACAAG[A/C]TTTTATTGGTGTCAGCATCTTTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605687 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL19 - interleukin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013371.3 | Intron | NP_037503.2 | ||||
NM_153758.2 | Intron | NP_715639.1 | ||||
XM_011509450.2 | Intron | XP_011507752.1 |