Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTGTCTCTATAAAGGCTTCTCC[A/G]TCCCACCTGAGTCAGACCCAAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605687 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL19 - interleukin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013371.3 | Intron | NP_037503.2 | ||||
NM_153758.2 | Intron | NP_715639.1 | ||||
XM_011509450.2 | Intron | XP_011507752.1 |