Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGGAGGGGCTTCAAGGGAAC[G/T]TGGGGAGCCAGGACAGGCAGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605048 MIM: 606524 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKBKE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IKBKE - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193321.1 | Intron | NP_001180250.1 | ||||
NM_001193322.1 | Intron | NP_001180251.1 | ||||
NM_014002.3 | Intron | NP_054721.1 | ||||
XM_005273356.2 | Intron | XP_005273413.1 | ||||
XM_017002868.1 | Intron | XP_016858357.1 |
MIR6769B - microRNA 6769b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRGAP2 - SLIT-ROBO Rho GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |