Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGAAGAACAAAACCTGTAATG[A/T]GTTTGCGAATATTAGTACTGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300086 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPAR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LPAR4 - lysophosphatidic acid receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278000.1 | 752 | Intron | NP_001264929.1 | |||
NM_005296.2 | 752 | Intron | NP_005287.1 | |||
XM_005262126.3 | 752 | Intron | XP_005262183.1 | |||
XM_006724639.3 | 752 | Intron | XP_006724702.1 | |||
XM_017029437.1 | 752 | UTR 5 | XP_016884926.1 | |||
XM_017029438.1 | 752 | Intron | XP_016884927.1 |
Set Membership: |
HapMap |