Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 610627 | |||||||||||||||||||||||||||||
Literature Links: |
A2ML1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
A2ML1 - alpha-2-macroglobulin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282424.2 | Intron | NP_001269353.1 | ||||
NM_144670.5 | Intron | NP_653271.2 | ||||
XM_011520566.2 | Intron | XP_011518868.1 | ||||
XM_011520567.2 | Intron | XP_011518869.1 | ||||
XM_017018868.1 | Intron | XP_016874357.1 | ||||
XM_017018869.1 | Intron | XP_016874358.1 | ||||
XM_017018870.1 | Intron | XP_016874359.1 |
Set Membership: |
HapMap |