Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCAGAAAATTTCTGAGCACTGA[C/T]GTGATGCCAGAAGTGGAAAATTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600392 MIM: 605232 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAD52 - RAD52 homolog, DNA repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297419.1 | 2828 | UTR 3 | NP_001284348.1 | |||
NM_001297420.1 | 2828 | Intron | NP_001284349.1 | |||
NM_001297421.1 | 2828 | UTR 3 | NP_001284350.1 | |||
NM_001297422.1 | 2828 | Intron | NP_001284351.1 | |||
NM_134424.3 | 2828 | UTR 3 | NP_602296.2 | |||
XM_005253720.4 | 2828 | UTR 3 | XP_005253777.1 | |||
XM_005253721.2 | 2828 | UTR 3 | XP_005253778.1 | |||
XM_011520990.2 | 2828 | UTR 3 | XP_011519292.1 | |||
XM_011520991.2 | 2828 | UTR 3 | XP_011519293.1 | |||
XM_011520993.2 | 2828 | Intron | XP_011519295.1 | |||
XM_011520995.2 | 2828 | UTR 3 | XP_011519297.1 | |||
XM_017019769.1 | 2828 | UTR 3 | XP_016875258.1 | |||
XM_017019770.1 | 2828 | UTR 3 | XP_016875259.1 | |||
XM_017019771.1 | 2828 | Intron | XP_016875260.1 |
Set Membership: |
HapMap |