Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616102 | ||||||||||||||||||||
Literature Links: |
CCDC102B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC102B - coiled-coil domain containing 102B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093729.1 | Intron | NP_001087198.1 | ||||
NM_024781.2 | Intron | NP_079057.2 | ||||
XM_011526174.2 | Intron | XP_011524476.1 | ||||
XM_011526175.2 | Intron | XP_011524477.1 | ||||
XM_017025973.1 | Intron | XP_016881462.1 | ||||
XM_017025974.1 | Intron | XP_016881463.1 | ||||
XM_017025975.1 | Intron | XP_016881464.1 | ||||
XM_017025976.1 | Intron | XP_016881465.1 | ||||
XM_017025977.1 | Intron | XP_016881466.1 | ||||
XM_017025978.1 | Intron | XP_016881467.1 | ||||
XM_017025979.1 | Intron | XP_016881468.1 | ||||
XM_017025980.1 | Intron | XP_016881469.1 |
TMX3 - thioredoxin related transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |