Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGAGTACAGAGGCCGCTCGCCC[A/G]ACGCCCACAGCGGGGGCCGCAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRCH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRCH2 - leucine rich repeats and calponin homology domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243963.1 | 1234 | Intron | NP_001230892.1 | |||
NM_020871.3 | 1234 | Intron | NP_065922.3 | |||
XM_006724724.3 | 1234 | Intron | XP_006724787.2 | |||
XM_017029696.1 | 1234 | Intron | XP_016885185.1 | |||
XM_017029697.1 | 1234 | Intron | XP_016885186.1 |
RBMXL3 - RNA binding motif protein, X-linked like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145346.1 | 1234 | Missense Mutation | AAC,GAC | N,D 398 | NP_001138818.1 |