Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATAACAGCAATTCTAACTTCAT[A/G]GTTTTCTCATGGTAATTAAATGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609874 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITLN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITLN2 - intelectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080878.2 | Intron | NP_543154.1 | ||||
XM_011509210.1 | Intron | XP_011507512.1 | ||||
XM_011509211.1 | Intron | XP_011507513.1 |
LOC101928372 - uncharacterized LOC101928372 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |