Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCAAAGCTGGGCAGATTTGCT[A/G]TCTCAGACCCGTCCCATCTTCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611930 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISG20L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ISG20L2 - interferon stimulated exonuclease gene 20 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303095.1 | Intron | NP_001290024.1 | ||||
XM_005245516.4 | Intron | XP_005245573.1 | ||||
XM_005245518.4 | Intron | XP_005245575.1 | ||||
XM_006711560.3 | Intron | XP_006711623.1 | ||||
XM_006711561.3 | Intron | XP_006711624.1 |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |