Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTCTTTAATGCTATCTTCTGC[C/T]GACACTAATCAAACTTCTTGAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA5 - proteasome subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199772.1 | Intron | NP_001186701.1 | ||||
NM_001199773.1 | Intron | NP_001186702.1 | ||||
NM_001199774.1 | Intron | NP_001186703.1 | ||||
NM_002790.3 | Intron | NP_002781.2 |