Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGCCAGCATGTGGCAACACTA[A/G]AATAAATTGGCAGACTCAACACCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182310 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP1A1 - ATPase Na+/K+ transporting subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000701.7 | 2278 | Intron | NP_000692.2 | |||
NM_001160233.1 | 2278 | Intron | NP_001153705.1 | |||
NM_001160234.1 | 2278 | Intron | NP_001153706.1 | |||
XM_017001360.1 | 2278 | UTR 5 | XP_016856849.1 | |||
XM_017001361.1 | 2278 | UTR 5 | XP_016856850.1 |
Set Membership: |
HapMap |