Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTAATCCCCATACCCCATAAGC[C/G]CCAGAGCCTCCATTCTGTTGCTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153420 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144822.1 | 18317 | Intron | NP_001138294.1 | |||
NM_001779.2 | 18317 | Intron | NP_001770.1 | |||
XM_017002869.1 | 18317 | UTR 3 | XP_016858358.1 |
LOC101929023 - uncharacterized LOC101929023 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378922 - uncharacterized LOC105378922 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |