Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGTCAATTATATGAACTAATAT[A/G]TCCAATCCCACCATATCTCAAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153420 MIM: 603491 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGSF3 - immunoglobulin superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007237.2 | Intron | NP_001007238.1 | ||||
NM_001542.3 | Intron | NP_001533.2 | ||||
XM_005270793.2 | Intron | XP_005270850.1 | ||||
XM_005270794.4 | Intron | XP_005270851.1 | ||||
XM_006710593.3 | Intron | XP_006710656.1 | ||||
XM_011541315.1 | Intron | XP_011539617.1 | ||||
XM_011541316.2 | Intron | XP_011539618.1 |
Set Membership: |
HapMap |