Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGAGGCAGGCCTCAAGGAGTA[G/T]CCAGGTAGGGTTACCTTTCCACCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PLA2G4E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLA2G4E - phospholipase A2 group IVE | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206670.1 | Intron | NP_001193599.1 | ||||
XM_011521237.2 | Intron | XP_011519539.1 | ||||
XM_011521238.1 | Intron | XP_011519540.1 | ||||
XM_011521239.1 | Intron | XP_011519541.1 |
PLA2G4E-AS1 - PLA2G4E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |