Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCGGGAGAACATCGAGGGACC[A/G]GTGGAGGAAGGGTAGCTGGATGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602534 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNAP23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNAP23 - synaptosome associated protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003825.3 | Intron | NP_003816.2 | ||||
NM_130798.2 | Intron | NP_570710.1 | ||||
XM_006720725.2 | Intron | XP_006720788.1 | ||||
XM_017022692.1 | Intron | XP_016878181.1 | ||||
XM_017022693.1 | Intron | XP_016878182.1 | ||||
XM_017022694.1 | Intron | XP_016878183.1 |
ZNF106 - zinc finger protein 106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |