Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAACCTGGACAGTCATTGCCTA[A/G]GATTCTATATAACTTTGAATAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614642 | ||||||||||||||||||||
Literature Links: |
STARD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STARD9 - StAR related lipid transfer domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020759.2 | Intron | NP_065810.2 | ||||
XM_011521831.2 | Intron | XP_011520133.1 | ||||
XM_011521832.2 | Intron | XP_011520134.1 | ||||
XM_011521833.2 | Intron | XP_011520135.1 | ||||
XM_011521834.2 | Intron | XP_011520136.1 | ||||
XM_011521835.2 | Intron | XP_011520137.1 | ||||
XM_011521836.2 | Intron | XP_011520138.1 | ||||
XM_011521837.2 | Intron | XP_011520139.1 | ||||
XM_011521839.2 | Intron | XP_011520141.1 | ||||
XM_017022439.1 | Intron | XP_016877928.1 | ||||
XM_017022440.1 | Intron | XP_016877929.1 | ||||
XM_017022441.1 | Intron | XP_016877930.1 | ||||
XM_017022442.1 | Intron | XP_016877931.1 | ||||
XM_017022443.1 | Intron | XP_016877932.1 |