Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACCACTTATCTAATTGATGTC[A/C]ACTTTGGGAGTGGATTATTAAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609173 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KNL1 - kinetochore scaffold 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144508.4 | Intron | NP_653091.3 | ||||
NM_170589.4 | Intron | NP_733468.3 | ||||
XM_017022432.1 | Intron | XP_016877921.1 |