Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGAGTGTTTCTGTTGTAAGCA[C/T]GAGTTGGAAGGGACAGATAAGTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614587 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHAC1 - ChaC glutathione specific gamma-glutamylcyclotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142776.1 | Intron | NP_001136248.1 | ||||
NM_024111.3 | Intron | NP_077016.2 | ||||
XM_017022563.1 | Intron | XP_016878052.1 | ||||
XM_017022564.1 | Intron | XP_016878053.1 | ||||
XM_017022565.1 | Intron | XP_016878054.1 |