Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATATGAATCTTCAAGAGCTGTT[C/G]AGATGGAAAATGCATTTGTCAGAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM182 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM182 - transmembrane protein 182 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321343.1 | Intron | NP_001308272.1 | ||||
NM_001321344.1 | Intron | NP_001308273.1 | ||||
NM_001321345.1 | Intron | NP_001308274.1 | ||||
NM_001321346.1 | Intron | NP_001308275.1 | ||||
NM_144632.4 | Intron | NP_653233.4 | ||||
XM_006712287.1 | Intron | XP_006712350.1 | ||||
XM_006712288.3 | Intron | XP_006712351.1 | ||||
XM_011510632.2 | Intron | XP_011508934.1 | ||||
XM_017003375.1 | Intron | XP_016858864.1 | ||||
XM_017003376.1 | Intron | XP_016858865.1 |