Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTTCCAGGAGCAGGCTGTGGCA[A/G]CCCCAGCATCTAAGTGGTTTAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD9 - major facilitator superfamily domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322080.1 | 2343 | UTR 3 | NP_001309009.1 | |||
NM_001322081.1 | 2343 | UTR 3 | NP_001309010.1 | |||
NM_032718.4 | 2343 | UTR 3 | NP_116107.3 | |||
XM_011512005.2 | 2343 | UTR 3 | XP_011510307.1 | |||
XM_011512006.2 | 2343 | UTR 3 | XP_011510308.1 | |||
XM_011512007.2 | 2343 | UTR 3 | XP_011510309.1 |
SLC9A2 - solute carrier family 9 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |