Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATATTAAGATGTGTTCTAGCATA[C/T]TGGAGATGACATTTTTCAAAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600530 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MFSD9 - major facilitator superfamily domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322080.1 | Intron | NP_001309009.1 | ||||
NM_001322081.1 | Intron | NP_001309010.1 | ||||
NM_032718.4 | Intron | NP_116107.3 | ||||
XM_011512005.2 | Intron | XP_011510307.1 | ||||
XM_011512006.2 | Intron | XP_011510308.1 | ||||
XM_011512007.2 | Intron | XP_011510309.1 |
SLC9A2 - solute carrier family 9 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |