Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTACAAAGGCAAGGTCAACGAGA[C/T]TGGGTGGTACTGGCATAAGGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607758 | ||||||||||||||||||||
Literature Links: |
CTNNBIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTNNBIP1 - catenin beta interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012329.1 | Intron | NP_001012329.1 | ||||
NM_020248.2 | Intron | NP_064633.1 | ||||
XM_017001853.1 | Intron | XP_016857342.1 | ||||
XM_017001854.1 | Intron | XP_016857343.1 | ||||
XM_017001855.1 | Intron | XP_016857344.1 |