Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACATTACCACGACGTATCTCAC[A/C]CACATTAATGCAGTCACGTACATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601791 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004565.2 | 433 | Intron | NP_004556.1 | |||
XM_005263470.4 | 433 | Intron | XP_005263527.1 | |||
XM_011541577.2 | 433 | Intron | XP_011539879.1 | |||
XM_011541578.2 | 433 | Intron | XP_011539880.1 | |||
XM_011541579.2 | 433 | UTR 5 | XP_011539881.1 | |||
XM_011541580.1 | 433 | Intron | XP_011539882.1 |
Set Membership: |
HapMap |